Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDLRAD1
Entrez Gene ID
388633
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 54007298 - 54018186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010978.3 NP_001010978.2 3 - 4 276 70
XM_011541446.1 XP_011539748.1 3 - 4 256 70
GenBank mRNA BC127708.1 - 3 - 4 273 70
CF887342.1 - 3 - 4 276 70

View Full Product Details