The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022551.2 | NP_072045.1 | 4 - 5 | 341 | 93 |
GenBank mRNA | AY536376.1 | - | 5 - 6 | 478 | 93 |
BC101786.1 | - | 4 - 5 | 336 | 93 | |
BC101788.1 | - | 4 - 5 | 336 | 93 | |
BC106063.1 | - | 4 - 5 | 394 | 93 | |
X69150.1 | - | 4 - 5 | 342 | 93 |