The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278641.1 | NP_001265570.1 | 1 - 2 | 355 | 73 |
XM_011530291.2 | XP_011528593.1 | 1 - 2 | 549 | 73 | |
XM_017028891.1 | XP_016884380.1 | 1 - 2 | 350 | 73 | |
GenBank mRNA | BG717186.1 | - | 1 - 2 | 104 | 73 |
BI916757.1 | - | 1 - 2 | 355 | 73 |