Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIXDC1
Entrez Gene ID
85458
Gene Name
DIX domain containing 1
Gene Aliases
CCD1
Chromosome Location
Chr.11: 111927144 - 112022650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278542.1 NP_001265471.1 2 - 3 332 69
XM_017018466.1 XP_016873955.1 2 - 3 312 69
XM_017018467.1 XP_016873956.1 2 - 3 180 69
GenBank mRNA BC048294.1 - 1 - 2 45 69
DB459100.1 - 2 - 3 334 69
HY037299.1 - 2 - 3 314 69

View Full Product Details