The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037954.3 | NP_001033043.1 | 4 - 5 | 850 | 80 |
NM_001278542.1 | NP_001265471.1 | 5 - 6 | 830 | 80 | |
XM_017018466.1 | XP_016873955.1 | 5 - 6 | 810 | 80 | |
XM_017018467.1 | XP_016873956.1 | 5 - 6 | 678 | 80 | |
GenBank mRNA | BC035509.2 | - | 4 - 5 | 666 | 80 |
BC048294.1 | - | 4 - 5 | 543 | 80 | |
BC064479.1 | - | 4 - 5 | 666 | 80 | |
BC128600.1 | - | 4 - 5 | 626 | 80 | |
DQ642016.1 | - | 4 - 5 | 850 | 80 |