The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_133652.1 | 13 - 14 | 2872 | 72 | |
XM_011516065.1 | XP_011514367.1 | 13 - 14 | 2872 | 72 | |
GenBank mRNA | AY316116.1 | - | 8 - 9 | 1210 | 72 |
AY515311.1 | - | 13 - 14 | 2872 | 72 |