Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSMD3
Entrez Gene ID
114788
Gene Name
CUB and Sushi multiple domains 3
Gene Aliases
-
Chromosome Location
Chr.8: 112222928 - 113437499 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052900.2 NP_443132.3 43 - 44 6864 105
NM_198123.1 NP_937756.1 44 - 45 7176 105
NM_198124.1 NP_937757.1 45 - 46 7068 105
XM_011516815.2 XP_011515117.1 18 - 19 2677 105
XM_011516816.2 XP_011515118.1 43 - 44 7350 105
XM_017013008.1 XP_016868497.1 43 - 44 7584 105
XM_017013009.1 XP_016868498.1 42 - 43 7452 105
XM_017013010.1 XP_016868499.1 42 - 43 7272 105
XM_017013011.1 XP_016868500.1 40 - 41 7062 105
XM_017013012.1 XP_016868501.1 42 - 43 6635 105
GenBank mRNA AB067481.2 - 29 - 30 4827 105
AB114604.1 - 44 - 45 7176 105
AB114605.1 - 45 - 46 7068 105
AK126252.1 - 2 - 3 209 105
AY210419.1 - 43 - 44 6735 105

View Full Product Details