The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321729.1 | NP_001308658.1 | 20 - 21 | 2324 | 84 |
NM_001321730.1 | NP_001308659.1 | 19 - 20 | 2189 | 84 | |
NM_001321731.1 | NP_001308660.1 | 20 - 21 | 2324 | 84 | |
NM_001321733.1 | NP_001308662.1 | 19 - 20 | 2189 | 84 | |
NM_001321734.1 | NP_001308663.1 | 19 - 20 | 2158 | 84 | |
NM_015189.2 | NP_056004.1 | 20 - 21 | 2324 | 84 | |
NR_135774.1 | 18 - 19 | 2002 | 84 | ||
XM_005264224.1 | XP_005264281.1 | 13 - 14 | 1616 | 84 | |
XM_017003641.1 | XP_016859130.1 | 16 - 17 | 1758 | 84 | |
XM_017003642.1 | XP_016859131.1 | 14 - 15 | 1371 | 84 | |
GenBank mRNA | AK023791.1 | - | 8 - 9 | 885 | 84 |
AK026474.1 | - | 17 - 18 | 1819 | 84 |