The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005140.1 | NP_005131.1 | 5 - 6 | 707 | 77 |
GenBank mRNA | AK128186.1 | - | 5 - 6 | 707 | 77 |
BC126302.1 | - | 5 - 6 | 576 | 77 | |
BC126304.1 | - | 5 - 6 | 576 | 77 |