The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100388.1 | NP_001093858.1 | 2 - 3 | 263 | 61 |
NM_207430.2 | NP_997313.2 | 2 - 3 | 263 | 61 | |
GenBank mRNA | AK128145.1 | - | 2 - 3 | 263 | 61 |
BC039505.2 | - | 2 - 3 | 248 | 61 |