Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RANBP1
Entrez Gene ID
5902
Gene Name
RAN binding protein 1
Gene Aliases
HTF9A
Chromosome Location
Chr.22: 20115938 - 20127357 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK097659.1 - 3 - 4 756 63

View Full Product Details