Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUTM2B
Entrez Gene ID
729262
Gene Name
NUT family member 2B
Gene Aliases
FAM22B, bA119F19.1
Chromosome Location
Chr.10: 79703227 - 79712757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278495.1 NP_001265424.1 4 - 5 1732 81

Gene Symbol
NUTM2A
Entrez Gene ID
728118
Gene Name
NUT family member 2A
Gene Aliases
FAM22A
Chromosome Location
Chr.10: 87221899 - 87235372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099338.1 NP_001092808.1 4 - 5 1732 81
XM_011540122.2 XP_011538424.1 6 - 7 4575 81
XM_017016607.1 XP_016872096.1 6 - 7 4576 81
GenBank mRNA BC092519.1 - 4 - 5 1728 81

Gene Symbol
NUTM2D
Entrez Gene ID
728130
Gene Name
NUT family member 2D
Gene Aliases
FAM22D
Chromosome Location
Chr.10: 87357720 - 87370695 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_075100.1 4 - 5 1679 81
GenBank mRNA AB095940.1 - 4 - 5 1679 81

View Full Product Details