The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080471.1 | NP_001073940.1 | 15 - 16 | 2070 | 88 |
XM_005245141.3 | XP_005245198.1 | 16 - 17 | 2263 | 88 | |
XM_011509510.2 | XP_011507812.1 | 16 - 17 | 2202 | 88 | |
XM_011509511.2 | XP_011507813.1 | 15 - 16 | 2158 | 88 | |
XM_011509512.2 | XP_011507814.1 | 15 - 16 | 2082 | 88 | |
XM_017001234.1 | XP_016856723.1 | 15 - 16 | 3328 | 88 | |
XM_017001235.1 | XP_016856724.1 | 15 - 16 | 3198 | 88 | |
XM_017001236.1 | XP_016856725.1 | 15 - 16 | 2165 | 88 | |
XM_017001237.1 | XP_016856726.1 | 16 - 17 | 2409 | 88 | |
XM_017001238.1 | XP_016856727.1 | 16 - 17 | 2348 | 88 | |
XM_017001239.1 | XP_016856728.1 | 16 - 17 | 2159 | 88 | |
XM_017001240.1 | XP_016856729.1 | 13 - 14 | 1727 | 88 | |
XM_017001241.1 | XP_016856730.1 | 13 - 14 | 1875 | 88 | |
XM_017001242.1 | XP_016856731.1 | 13 - 14 | 1779 | 88 | |
GenBank mRNA | AK074121.1 | - | 10 - 11 | 1682 | 88 |