The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198486.3 | NP_940888.3 | 4 - 5 | 743 | 88 |
NR_134562.1 | 4 - 5 | 859 | 88 | ||
NR_134563.1 | 3 - 4 | 637 | 88 | ||
XM_017010780.1 | XP_016866269.1 | 4 - 5 | 1196 | 88 | |
GenBank mRNA | AK021904.1 | - | 3 - 4 | 324 | 88 |
AK291310.1 | - | 3 - 4 | 346 | 88 | |
AK291949.1 | - | 4 - 5 | 430 | 88 | |
BC058020.1 | - | 4 - 5 | 434 | 88 | |
BC073890.1 | - | 4 - 5 | 426 | 88 |