The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165030.2 | NP_001158502.1 | 2 - 3 | 561 | 110 |
NM_015012.3 | NP_055827.1 | 2 - 3 | 561 | 110 | |
NR_028491.2 | 2 - 3 | 561 | 110 | ||
GenBank mRNA | AK303339.1 | - | 2 - 3 | 252 | 110 |
BC035034.1 | - | 2 - 3 | 432 | 110 | |
BC044597.1 | - | 2 - 3 | 423 | 110 | |
BC064366.1 | - | 2 - 3 | 318 | 110 | |
BC091524.1 | - | 2 - 3 | 360 | 110 | |
BE673436.1 | - | 1 - 2 | 110 | ||
BI596931.1 | - | 2 - 3 | 563 | 108 | |
BI829492.1 | - | 2 - 3 | 541 | 110 | |
BU752757.1 | - | 1 - 2 | 110 | ||
D26067.1 | - | 2 - 3 | 367 | 110 | |
DA149513.1 | - | 2 - 3 | 427 | 110 | |
DR157551.1 | - | 2 - 3 | 414 | 110 |