Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GVINP1
- Entrez Gene ID
-
387751
- Gene Name
- GTPase, very large interferon inducible pseudogene 1
- Gene Aliases
-
GVIN1, GVIN1P, VLIG-1, VLIG1
- Chromosome Location
-
Chr.11: 6713139 - 6721879 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX538318.1 |
- |
1 - 2 |
95 |
109 |
View Full Product Details