The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001124756.2 | NP_001118228.1 | 5 - 6 | 819 | 76 |
NR_134983.1 | 5 - 6 | 819 | 76 | ||
GenBank mRNA | EU190483.1 | - | 5 - 6 | 755 | 76 |