Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PABPC1L
- Entrez Gene ID
-
80336
- Gene Name
- poly(A) binding protein cytoplasmic 1 like
- Gene Aliases
-
C20orf119, EPAB, PABPC1L1, dJ1069P2.3
- Chromosome Location
-
Chr.20: 44910060 - 44959160 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_134987.1 |
|
3 - 4 |
1824 |
78 |
View Full Product Details