The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013661.1 | NP_001013683.1 | 3 - 4 | 568 | 64 |
GenBank mRNA | BC132893.1 | - | 3 - 4 | 460 | 64 |
BC132895.1 | - | 3 - 4 | 460 | 64 |