Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACCSL
Entrez Gene ID
390110
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
Gene Aliases
-
Chromosome Location
Chr.11: 43920987 - 44060667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031854.2 NP_001027025.2 12 - 13 1528 68
XM_017017714.1 XP_016873203.1 15 - 16 1901 68
XM_017017715.1 XP_016873204.1 12 - 13 1541 68
GenBank mRNA AB231734.1 - 12 - 13 1528 68
BC144554.1 - 12 - 13 1541 68
DW009984.1 - 2 - 3 227 68

View Full Product Details