The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144956.1 | NP_001138428.1 | 2 - 3 | 94 | |
GenBank mRNA | AA393535.1 | - | 2 - 3 | 94 | |
CK300240.1 | - | 3 - 4 | 316 | 94 |