The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_004844.1 | 1 - 1 | 664 | 107 | |
GenBank mRNA | BC067891.1 | - | 1 - 1 | 664 | 107 |