The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080464.2 | NP_001073933.2 | 15 - 16 | 1786 | 68 |
XM_017021267.1 | XP_016876756.1 | 14 - 15 | 2391 | 68 | |
XM_017021273.1 | XP_016876762.1 | 13 - 14 | 2253 | 68 | |
XM_017021276.1 | XP_016876765.1 | 12 - 13 | 2153 | 68 | |
GenBank mRNA | AW024833.1 | - | 1 - 2 | 68 | |
AX781433.1 | - | 15 - 16 | 1793 | 68 | |
BC136637.1 | - | 15 - 16 | 1728 | 68 |