The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277269.1 | NP_001264198.1 | 34 - 35 | 4388 | 73 |
NM_001292063.1 | NP_001278992.1 | 35 - 36 | 4352 | 73 | |
GenBank mRNA | AK128214.1 | - | 11 - 12 | 1690 | 73 |