Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GSTM2P1
- Entrez Gene ID
-
442245
- Gene Name
- glutathione S-transferase mu 2 pseudogene 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 111046419 - 111047554 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002932.2 |
|
1 - 1 |
635 |
65 |
GenBank mRNA |
AY532925.1 |
- |
1 - 1 |
602 |
65 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ribosome production factor 2 homolog |
RPF2 |
BXDC1, bA397G5.4 |
-18.2kb
|
Anti-sense |
Chr. - 110982017 - 111028263 on Build GRCh38 |
Downstream |
solute carrier family 16 member 10 |
SLC16A10 |
MCT10, PRO0813, TAT1 |
39.9kb
|
Anti-sense |
Chr. - 111087503 - 111227125 on Build GRCh38 |
View Full Product Details