Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC6A19
Entrez Gene ID
340024
Gene Name
solute carrier family 6 member 19
Gene Aliases
B0AT1, HND
Chromosome Location
Chr.5: 1201595 - 1225117 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003841.2 NP_001003841.1 2 - 3 394 73
GenBank mRNA AK290811.1 - 2 - 3 394 73
AY591756.1 - 2 - 3 338 73
AY596807.1 - 2 - 3 338 73

View Full Product Details