Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGK1
Entrez Gene ID
6446
Gene Name
serum/glucocorticoid regulated kinase 1
Gene Aliases
SGK
Chromosome Location
Chr.6: 134169246 - 134318112 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143676.1 NP_001137148.1 2 - 3 890 77
XM_011536071.1 XP_011534373.1 3 - 4 672 77
GenBank mRNA AK055077.1 - 2 - 3 890 77
EU518415.1 - 2 - 3 292 77
FM205709.1 - 2 - 3 739 77

View Full Product Details