The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010853.2 | NP_001010853.1 | 2 - 3 | 706 | 99 |
XM_005248661.2 | XP_005248718.1 | 3 - 4 | 705 | 99 | |
XM_005248662.4 | XP_005248719.1 | 3 - 4 | 456 | 99 | |
XM_011535481.2 | XP_011533783.1 | 2 - 3 | 938 | 99 | |
XM_017010289.1 | XP_016865778.1 | 2 - 3 | 706 | 99 | |
XM_017010290.1 | XP_016865779.1 | 2 - 3 | 528 | 99 | |
XM_017010291.1 | XP_016865780.1 | 3 - 4 | 348 | 99 | |
GenBank mRNA | AK302464.1 | - | 2 - 3 | 666 | 99 |
BC035036.1 | - | 2 - 3 | 709 | 99 | |
BC045583.1 | - | 2 - 3 | 457 | 99 |