Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSD2
Entrez Gene ID
123722
Gene Name
fibronectin type III and SPRY domain containing 2
Gene Aliases
SPRYD1
Chromosome Location
Chr.15: 82755365 - 82807698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007122.3 NP_001007123.1 5 - 6 1171 103
NM_001281805.1 NP_001268734.1 6 - 7 1288 103
NM_001281806.1 NP_001268735.1 5 - 6 1171 103
XM_005272425.4 XP_005272482.1 6 - 7 2914 103
XM_017021924.1 XP_016877413.1 6 - 7 2914 103
GenBank mRNA AK122875.1 - 5 - 6 1170 103
AL833295.1 - 5 - 6 103
BC130569.1 - 5 - 6 1155 103
BC144175.1 - 6 - 7 1272 103
BC144181.1 - 5 - 6 1155 103

View Full Product Details