Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC24A5
Entrez Gene ID
283652
Gene Name
solute carrier family 24 member 5
Gene Aliases
JSX, NCKX5, OCA6, SHEP4
Chromosome Location
Chr.15: 48120972 - 48142392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205850.2 NP_995322.1 3 - 4 457 79
GenBank mRNA AF348468.1 - 3 - 4 457 79
BC113628.1 - 3 - 4 412 79
BC113630.1 - 3 - 4 412 79
BC143950.1 - 3 - 4 412 79
DQ665306.1 - 3 - 4 405 79
DQ665307.1 - 2 - 3 225 79

View Full Product Details