Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC24A5
Entrez Gene ID
283652
Gene Name
solute carrier family 24 member 5
Gene Aliases
JSX, NCKX5, OCA6, SHEP4
Chromosome Location
Chr.15: 48120972 - 48142392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205850.2 NP_995322.1 6 - 7 950 95
XM_017022079.1 XP_016877568.1 5 - 6 2051 95
XM_017022080.1 XP_016877569.1 5 - 6 2271 95
GenBank mRNA AF348468.1 - 6 - 7 950 95
BC113628.1 - 6 - 7 905 95
BC113630.1 - 6 - 7 905 95
BC143950.1 - 6 - 7 905 95
DQ665306.1 - 6 - 7 898 95
DQ665307.1 - 5 - 6 718 95

View Full Product Details