The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013742.3 | NP_001013764.1 | 17 - 18 | 2608 | 97 |
XM_017029268.1 | XP_016884757.1 | 17 - 18 | 2720 | 97 | |
GenBank mRNA | AB183864.1 | - | 17 - 18 | 2608 | 97 |
BC137319.1 | - | 17 - 18 | 2592 | 97 | |
BC137320.1 | - | 17 - 18 | 2592 | 97 |