Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGKK
Entrez Gene ID
139189
Gene Name
diacylglycerol kinase kappa
Gene Aliases
-
Chromosome Location
Chr.X: 50365407 - 50470850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013742.3 NP_001013764.1 6 - 7 1238 99
XM_017029268.1 XP_016884757.1 6 - 7 1350 99
GenBank mRNA AB183864.1 - 6 - 7 1238 99
BC137319.1 - 6 - 7 1222 99
BC137320.1 - 6 - 7 1222 99

View Full Product Details