Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKK
Entrez Gene ID
139189
Gene Name
diacylglycerol kinase kappa
Gene Aliases
-
Chromosome Location
Chr.X: 50365407 - 50470850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013742.3 NP_001013764.1 9 - 10 1658 84
XM_017029268.1 XP_016884757.1 9 - 10 1770 84
GenBank mRNA AB183864.1 - 9 - 10 1658 84
BC137319.1 - 9 - 10 1642 84
BC137320.1 - 9 - 10 1642 84

View Full Product Details