The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145026.1 | NP_001138498.1 | 33 - 34 | 5690 | 159 |
XM_017019273.1 | XP_016874762.1 | 37 - 38 | 6492 | 159 | |
XM_017019274.1 | XP_016874763.1 | 37 - 38 | 6493 | 159 |