The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286615.1 | NP_001273544.1 | 2 - 3 | 625 | 79 |
NM_001286616.1 | NP_001273545.1 | 1 - 2 | 193 | 79 | |
XM_011537911.2 | XP_011536213.1 | 4 - 5 | 620 | 79 | |
XM_011537912.2 | XP_011536214.1 | 4 - 5 | 620 | 79 | |
XM_011537913.2 | XP_011536215.1 | 4 - 5 | 620 | 79 | |
XM_011537915.2 | XP_011536217.1 | 4 - 5 | 620 | 79 | |
XM_011537916.2 | XP_011536218.1 | 2 - 3 | 284 | 79 | |
XM_011537918.2 | XP_011536220.1 | 4 - 5 | 620 | 79 | |
XM_017018809.1 | XP_016874298.1 | 3 - 4 | 711 | 79 | |
XM_017018812.1 | XP_016874301.1 | 2 - 3 | 2523 | 79 | |
XM_017018814.1 | XP_016874303.1 | 2 - 3 | 2523 | 79 | |
XM_017018815.1 | XP_016874304.1 | 2 - 3 | 2523 | 79 | |
XM_017018817.1 | XP_016874306.1 | 2 - 3 | 2523 | 79 | |
GenBank mRNA | AK293810.1 | - | 2 - 3 | 263 | 79 |
AK302962.1 | - | 4 - 5 | 551 | 79 | |
AK316105.1 | - | 2 - 3 | 264 | 79 | |
BC109308.2 | - | 1 - 2 | 178 | 79 |