The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007137.3 | NP_009068.2 | 2 - 3 | 297 | 93 |
XM_005272600.3 | XP_005272657.1 | 2 - 3 | 355 | 93 | |
XM_011543899.2 | XP_011542201.1 | 2 - 3 | 1961 | 93 | |
XM_011543900.2 | XP_011542202.1 | 3 - 4 | 416 | 93 | |
XM_017029486.1 | XP_016884975.1 | 3 - 4 | 474 | 93 | |
XM_017029487.1 | XP_016884976.1 | 2 - 3 | 707 | 93 | |
GenBank mRNA | AK126949.1 | - | 3 - 4 | 416 | 93 |
BC039609.1 | - | 3 - 4 | 392 | 93 | |
DA219953.1 | - | 3 - 4 | 416 | 93 | |
DR002896.1 | - | 2 - 3 | 285 | 93 |