Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF81
Entrez Gene ID
347344
Gene Name
zinc finger protein 81
Gene Aliases
HFZ20, MRX45, dJ54B20.6
Chromosome Location
Chr.X: 47836864 - 47925626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007137.3 NP_009068.2 3 - 4 431 111
XM_005272600.3 XP_005272657.1 3 - 4 489 111
XM_011543899.2 XP_011542201.1 3 - 4 2095 111
XM_011543900.2 XP_011542202.1 4 - 5 550 111
XM_017029486.1 XP_016884975.1 4 - 5 608 111
XM_017029487.1 XP_016884976.1 3 - 4 841 111
GenBank mRNA AK126949.1 - 4 - 5 550 111
AY487248.1 - 2 - 3 182 111
DR002896.1 - 3 - 4 419 111

View Full Product Details