The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007137.3 | NP_009068.2 | 4 - 5 | 532 | 67 |
XM_005272600.3 | XP_005272657.1 | 4 - 5 | 590 | 67 | |
XM_011543899.2 | XP_011542201.1 | 4 - 5 | 2196 | 67 | |
XM_011543900.2 | XP_011542202.1 | 5 - 6 | 651 | 67 | |
XM_017029486.1 | XP_016884975.1 | 5 - 6 | 709 | 67 | |
XM_017029487.1 | XP_016884976.1 | 4 - 5 | 942 | 67 | |
GenBank mRNA | AK126949.1 | - | 5 - 6 | 651 | 67 |
AY487248.1 | - | 3 - 4 | 283 | 67 | |
DR002896.1 | - | 4 - 5 | 520 | 67 |