The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173591.3 | NP_775862.3 | 13 - 14 | 1377 | 110 |
XM_005268802.3 | XP_005268859.1 | 17 - 18 | 2178 | 110 | |
XM_011538191.1 | XP_011536493.1 | 14 - 15 | 1454 | 110 | |
XM_011538192.2 | XP_011536494.1 | 11 - 12 | 1395 | 110 | |
XM_011538193.1 | XP_011536495.1 | 11 - 12 | 1314 | 110 |