Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
Chromosome Location
Chr.12: 123761995 - 123935720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207437.3 NP_997320.2 16 - 17 2449 69
XM_005253555.3 XP_005253612.1 17 - 18 3112 69
XM_011538014.2 XP_011536316.1 17 - 18 3112 69
XM_011538015.2 XP_011536317.1 17 - 18 3112 69
XM_011538016.2 XP_011536318.1 17 - 18 3112 69
XM_011538017.2 XP_011536319.1 17 - 18 3112 69
XM_011538020.2 XP_011536322.1 17 - 18 3120 69
XM_017018960.1 XP_016874449.1 17 - 18 3112 69
XM_017018961.1 XP_016874450.1 17 - 18 3112 69
GenBank mRNA AK125475.1 - 11 - 12 1963 69
BC144575.1 - 12 - 13 2110 69
BC150622.1 - 12 - 13 2110 69

View Full Product Details