Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
Chromosome Location
Chr.12: 123761995 - 123935720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207437.3 NP_997320.2 26 - 27 4443 141
XM_005253555.3 XP_005253612.1 27 - 28 5106 141
XM_011538014.2 XP_011536316.1 27 - 28 5106 141
XM_011538015.2 XP_011536317.1 27 - 28 5106 141
XM_011538016.2 XP_011536318.1 27 - 28 5106 141
XM_011538017.2 XP_011536319.1 27 - 28 5106 141
XM_011538019.2 XP_011536321.1 9 - 10 1705 141
XM_011538020.2 XP_011536322.1 27 - 28 5114 141
XM_017018960.1 XP_016874449.1 27 - 28 5106 141
XM_017018961.1 XP_016874450.1 26 - 27 4797 141
XM_017018962.1 XP_016874451.1 9 - 10 1627 141
GenBank mRNA AB095937.2 - 1 - 2 158 141
AB290168.1 - 6 - 7 962 141

View Full Product Details