Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
Chromosome Location
Chr.12: 123761995 - 123935720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207437.3 NP_997320.2 38 - 39 6576 72
XM_005253555.3 XP_005253612.1 39 - 40 7239 72
XM_011538015.2 XP_011536317.1 38 - 39 7050 72
XM_011538016.2 XP_011536318.1 39 - 40 7239 72
XM_011538017.2 XP_011536319.1 37 - 38 6903 72
XM_011538019.2 XP_011536321.1 21 - 22 3838 72
XM_011538020.2 XP_011536322.1 39 - 40 7247 72
XM_017018960.1 XP_016874449.1 39 - 40 7239 72
XM_017018961.1 XP_016874450.1 38 - 39 6930 72
XM_017018962.1 XP_016874451.1 21 - 22 3760 72
GenBank mRNA AB095937.2 - 13 - 14 2291 72
AB290168.1 - 18 - 19 3095 72
AK125796.1 - 12 - 13 2108 72

View Full Product Details