Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
Chromosome Location
Chr.12: 123761995 - 123935720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207437.3 NP_997320.2 44 - 45 7444 80
XM_005253555.3 XP_005253612.1 45 - 46 8107 80
XM_011538014.2 XP_011536316.1 44 - 45 7948 80
XM_011538015.2 XP_011536317.1 44 - 45 7918 80
XM_011538016.2 XP_011536318.1 45 - 46 8107 80
XM_011538017.2 XP_011536319.1 43 - 44 7771 80
XM_011538019.2 XP_011536321.1 27 - 28 4706 80
XM_017018960.1 XP_016874449.1 45 - 46 8107 80
XM_017018961.1 XP_016874450.1 44 - 45 7798 80
XM_017018962.1 XP_016874451.1 27 - 28 4628 80
GenBank mRNA AB095937.2 - 19 - 20 3159 80
AB290168.1 - 24 - 25 3963 80

View Full Product Details