Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
Chromosome Location
Chr.12: 123761995 - 123935720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207437.3 NP_997320.2 49 - 50 8304 117
XM_005253555.3 XP_005253612.1 50 - 51 8967 117
XM_011538014.2 XP_011536316.1 49 - 50 8808 117
XM_011538015.2 XP_011536317.1 49 - 50 8778 117
XM_011538017.2 XP_011536319.1 48 - 49 8631 117
XM_011538019.2 XP_011536321.1 32 - 33 5566 117
XM_017018960.1 XP_016874449.1 50 - 51 8967 117
XM_017018961.1 XP_016874450.1 49 - 50 8658 117
XM_017018962.1 XP_016874451.1 32 - 33 5488 117
GenBank mRNA AB095937.2 - 24 - 25 4019 117
AB290168.1 - 29 - 30 4823 117
AK125796.1 - 20 - 21 3276 117

View Full Product Details