Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXXC1P1
Entrez Gene ID
392459
Gene Name
CXXC finger protein 1 pseudogene 1
Gene Aliases
CXorf25, NCRNA00236
Chromosome Location
Chr.X: 47707191 - 47736628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033924.1 2 - 3 1006 98
GenBank mRNA AK094108.1 - 2 - 3 1008 98

View Full Product Details