The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145112.1 | NP_001138584.1 | 9 - 10 | 968 | 71 |
XM_011521336.2 | XP_011519638.2 | 9 - 10 | 1794 | 71 | |
XM_011521337.2 | XP_011519639.2 | 9 - 10 | 1929 | 71 | |
XM_011521338.2 | XP_011519640.1 | 11 - 12 | 1493 | 71 | |
XM_011521339.2 | XP_011519641.1 | 10 - 11 | 1374 | 71 | |
XM_011521340.2 | XP_011519642.1 | 9 - 10 | 1315 | 71 | |
XM_011521341.1 | XP_011519643.1 | 9 - 10 | 1078 | 71 | |
XM_011521342.2 | XP_011519644.1 | 7 - 8 | 1975 | 71 | |
XM_011521343.2 | XP_011519645.1 | 10 - 11 | 1325 | 71 | |
XM_011521344.2 | XP_011519646.1 | 7 - 8 | 767 | 71 | |
XM_011521345.2 | XP_011519647.1 | 10 - 11 | 1287 | 71 | |
XM_011521346.2 | XP_011519648.2 | 5 - 6 | 1501 | 71 | |
XM_011521347.1 | XP_011519649.1 | 8 - 9 | 883 | 71 | |
XM_011521348.2 | XP_011519650.1 | 8 - 9 | 883 | 71 | |
XM_017022000.1 | XP_016877489.1 | 9 - 10 | 1786 | 71 | |
XM_017022001.1 | XP_016877490.1 | 7 - 8 | 915 | 71 | |
GenBank mRNA | BI909158.1 | - | 1 - 2 | 68 | 71 |
DR007980.1 | - | 1 - 2 | 71 |