The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040709.1 | NP_001035799.1 | 4 - 5 | 823 | 78 |
XM_011541283.2 | XP_011539585.1 | 4 - 5 | 823 | 78 | |
GenBank mRNA | AB158472.1 | - | 4 - 5 | 457 | 78 |
AK131459.1 | - | 4 - 5 | 823 | 78 | |
AK289513.1 | - | 4 - 5 | 757 | 78 | |
AK293082.1 | - | 4 - 5 | 699 | 78 | |
AK301509.1 | - | 1 - 2 | 81 | 78 | |
BC113102.1 | - | 1 - 2 | 232 | 78 | |
BC113103.1 | - | 1 - 2 | 220 | 78 | |
DN831287.1 | - | 1 - 2 | 215 | 78 |