The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080442.2 | NP_001073911.1 | 9 - 10 | 1219 | 70 |
XM_017022946.1 | XP_016878435.1 | 11 - 12 | 1684 | 70 |