The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042367.1 | NP_001035826.1 | 2 - 3 | 282 | 140 |
GenBank mRNA | BC055412.1 | - | 2 - 3 | 263 | 140 |